Assay Details
Target Gene Details
Entrez Gene ID: | 116442 |
Gene Name: | RAB39B, member RAS oncogene family |
Gene Aliases: |
MRX72, WSMN |
Location: |
Chr.X:155258234-155264589 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB39B | NM_171998.3 | 2 | 2792 | NP_741995.1 |
| AK056209.1 | 1 | 1751 | ||
| AL834460.1 | 2 | 2655 | CAD39120.1 | |
| BX108796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map