Assay Details
Target Gene Details
Entrez Gene ID: | 1734 |
Gene Name: | deiodinase, iodothyronine, type II |
Gene Aliases: |
5DII, D2, DIOII, SelY, TXDI2 |
Location: |
Chr.14:80197525-80231054 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO2 | NM_000793.5 | 4 | 3348 | NP_000784.2 |
| NM_001007023.4 | 3 | 3225 | NP_001007024.1 | |
| NM_001242502.2 | 4 | 3359 | NP_001229431.1 | |
| NM_001242503.2 | 3 | 3251 | NP_001229432.1 | |
| NM_001324462.1 | 3 | 3364 | NP_001311391.1 | |
| NM_013989.4 | 2 | 3117 | NP_054644.1 | |
| AF093774.1 | 2 | 3117 | AAC95470.1 | |
| AF123661.1 | ||||
| AI701403.1 | ||||
| BC063118.1 | AAH63118.1 | |||
| BC136514.1 | 2 | 2987 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045241 | Chr.14:79419018 - 80267858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map