Assay Details
Target Gene Details
Entrez Gene ID: | 85450 |
Gene Name: | inositol 1,4,5-trisphosphate receptor interacting protein |
Gene Aliases: |
DANGER, KIAA1754, bA127L20, bA127L20.2 |
Location: |
Chr.10:104309696-104338493 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPRIP | NM_001272012.1 | 2 | 1445 | NP_001258941.1 |
| NM_001272013.1 | 2 | 1565 | NP_001258942.1 | |
| NM_033397.3 | 3 | 1757 | NP_203755.1 | |
| XM_005270257.2 | 1 | 1321 | XP_005270314.1 | |
| AB051541.1 | 2 | 1476 | ||
| AK055608.1 | 1 | 168 | ||
| AK056526.1 | 2 | 1377 | BAB71204.1 | |
| AK094802.1 | 1 | 797 | ||
| AK129877.1 | 3 | 1468 | ||
| BC040536.1 | 2 | 1417 | AAH40536.1 | |
| BC070108.1 | 3 | 1757 | AAH70108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951841 | Chr.10:104311343 - 104316942 on Build GRCh38 | Deletion |
|

Genomic Map