Assay Details
Target Gene Details
Entrez Gene ID: | 6950 |
Gene Name: | t-complex 1 |
Gene Aliases: |
CCT-alpha, CCT1, CCTa, D6S230E, TCP-1-alpha |
Location: |
Chr.6:159778498-159789703 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCP1 | NM_001008897.1 | NP_001008897.1 | ||
| NM_030752.2 | NP_110379.2 | |||
| AK312604.1 | ||||
| BC000665.1 | AAH00665.1 | |||
| BT006969.1 | AAP35615.1 | |||
| X52882.1 | CAA37064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5563 | Chr.6:159775783 - 159788607 on Build GRCh38 | Insertion |
|
| esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map