Assay Details
Target Gene Details
Entrez Gene ID: | 79814 |
Gene Name: | agmatinase |
Gene Aliases: |
- |
Location: |
Chr.1:15571699-15585110 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGMAT | NM_024758.4 | NP_079034.3 | ||
| AK130034.1 | ||||
| AK225366.1 | ||||
| AY057097.1 | AAL24446.1 | |||
| BC005090.1 | AAH05090.1 |
More Information
Set Membership: |
|

Genomic Map