Assay Details
Target Gene Details
Entrez Gene ID: | 29902 |
Gene Name: | family with sequence similarity 216 member A |
Gene Aliases: |
C12orf24, HSU79274 |
Location: |
Chr.12:110468424-110490387 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM216A | NM_013300.2 | NP_037432.2 | ||
| XM_005253875.4 | XP_005253932.1 | |||
| XM_011538246.2 | XP_011536548.1 | |||
| AK225731.1 | ||||
| AK296280.1 | ||||
| AK297684.1 | ||||
| AK308802.1 | ||||
| AK309603.1 | ||||
| BC020967.1 | AAH20967.1 | |||
| CR457001.1 | CAG33282.1 | |||
| U79274.1 | AAB50215.1 |

Genomic Map