Assay Details
Target Gene Details
Entrez Gene ID: | 3738 |
Gene Name: | potassium voltage-gated channel subfamily A member 3 |
Gene Aliases: |
HGK5, HLK3, HPCN3, HUKIII, KV1.3, MK3, PCN3 |
Location: |
Chr.1:110653560-110675033 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA3 | NM_002232.4 | 1 | 1103 | NP_002223.3 |
| AK314009.1 | 1 | 1010 | ||
| BC035059.1 | 1 | 1104 | AAH35059.2 | |
| BC064595.1 | 1 | 826 | AAH64595.1 | |
| L23499.1 | 1 | 895 | AAC31761.1 | |
| M85217.1 | 1 | 810 | AAA59457.1 |
More Information
Set Membership: |
|

Genomic Map