Assay Details
Target Gene Details
Entrez Gene ID: | 54854 |
Gene Name: | family with sequence similarity 83 member E |
Gene Aliases: |
- |
Location: |
Chr.19:48600600-48614841 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM83E | NM_017708.3 | NP_060178.2 | ||
| XM_005259019.1 | XP_005259076.1 | |||
| XM_011527049.2 | XP_011525351.1 | |||
| AK000207.1 | BAA91009.1 | |||
| BC111970.1 | AAI11971.1 | |||
| BC111972.1 | AAI11973.1 |
Target Gene Details
Entrez Gene ID: | 171169 |
Gene Name: | sperm acrosome associated 4 |
Gene Aliases: |
SAMP14 |
Location: |
Chr.19:48606743-48607714 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPACA4 | NM_133498.2 | 1 | 365 | NP_598005.1 |
| AF353721.2 | 1 | 365 | AAL83950.1 | |
| AY358387.1 | 1 | 347 | AAQ88753.1 | |
| BC047334.1 | 1 | 345 | AAH47334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv666e214 | Chr.19:48604616 - 48609671 on Build GRCh38 | Loss |
|
| dgv3617n100 | Chr.19:48532114 - 48643732 on Build GRCh38 | Gain |
|
| nsv1057580 | Chr.19:48512123 - 48608737 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map