Assay Details
Target Gene Details
Entrez Gene ID: | 55466 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A4 |
Gene Aliases: |
MST104, MSTP104, PRO1472 |
Location: |
Chr.15:78264145-78282196 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA4 | NM_001130182.1 | 7 | 1168 | NP_001123654.1 |
| NM_001130183.1 | 7 | 995 | NP_001123655.1 | |
| NM_018602.3 | 8 | 1307 | NP_061072.3 | |
| XM_005254535.3 | 8 | 1080 | XP_005254592.1 | |
| XM_006720601.1 | 8 | 1082 | XP_006720664.1 | |
| AF173388.1 | 3 | 906 | AAQ13629.1 | |
| AK025069.1 | 2 | 290 | ||
| AK090898.1 | 4 | 639 | BAC03540.1 | |
| AK096616.1 | 7 | 1168 | BAC04828.1 | |
| AK098079.1 | 8 | 1298 | BAC05229.1 | |
| AL133096.1 | 2 | 1080 | CAH10713.1 | |
| AL133431.1 | ||||
| AL133433.1 | 2 | 409 | ||
| AL133434.1 | 2 | 506 | ||
| BC021720.2 | 8 | 1307 | AAH21720.1 | |
| BC031044.1 | 7 | 1030 | AAH31044.2 | |
| BX648710.1 | 8 | 1189 | CAH10558.1 |
More Information
Set Membership: |
|

Genomic Map