Assay Details
Target Gene Details
Entrez Gene ID: | 8390 |
Gene Name: | olfactory receptor family 1 subfamily G member 1 |
Gene Aliases: |
OR17-130, OR17-209, OR1G2 |
Location: |
Chr.17:3126610-3127551 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1G1 | NM_003555.1 | 1 | 324 | NP_003546.1 |
| BC069379.1 | 1 | 324 | AAH69379.1 | |
| BC095520.1 | 1 | 355 | AAH95520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| dgv3084n100 | Chr.17:3045742 - 3209683 on Build GRCh38 | Loss |
|
| esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion |
|
| nsv1060926 | Chr.17:3093486 - 3319621 on Build GRCh38 | Gain |
|
| dgv4n29 | Chr.17:3086643 - 3256571 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79847161] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map