Assay Details
Target Gene Details
Entrez Gene ID: | 26057 |
Gene Name: | ankyrin repeat domain 17 |
Gene Aliases: |
GTAR, MASK2, NY-BR-16 |
Location: |
Chr.4:73073966-73259142 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD17 | NM_001286771.2 | 29 | 5958 | NP_001273700.1 |
| NM_015574.1 | 29 | 6337 | NP_056389.1 | |
| NM_032217.4 | 29 | 6340 | NP_115593.3 | |
| NM_198889.2 | 28 | 5587 | NP_942592.1 | |
| XM_005265671.3 | 28 | 5928 | XP_005265728.1 | |
| XM_017008011.1 | 29 | 5912 | XP_016863500.1 | |
| XM_017008012.1 | 28 | 5162 | XP_016863501.1 | |
| XM_017008013.1 | 28 | 5159 | XP_016863502.1 | |
| AB014597.2 | 29 | 5861 | BAA31672.2 | |
| AK096518.1 | 5 | 1684 | ||
| AL833287.1 | 25 | 4764 | ||
| BC019963.2 | 1 | 884 | AAH19963.1 | |
| BC043394.1 | AAH43394.1 | |||
| BX648035.1 | 29 | 5911 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600985 | Chr.4:72952867 - 73192514 on Build GRCh38 | Gain |
|
| nsv523169 | Chr.4:72886525 - 73255193 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map