Assay Details
Target Gene Details
Entrez Gene ID: | 11001 |
Gene Name: | solute carrier family 27 member 2 |
Gene Aliases: |
ACSVL1, FACVL1, FATP2, HsT17226, VLACS, VLCS, hFACVL1 |
Location: |
Chr.15:50182196-50236392 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC27A2 | NM_001159629.1 | 1 | 290 | NP_001153101.1 |
| NM_003645.3 | 1 | 290 | NP_003636.2 | |
| AF096290.1 | 1 | 262 | AAC64973.1 | |
| AK222859.1 | 1 | 261 | BAD96579.1 | |
| AK223145.1 | 1 | 259 | BAD96865.1 | |
| AK290262.1 | 1 | 290 | ||
| BC057770.1 | 1 | 253 | AAH57770.1 | |
| D88308.1 | 1 | 280 | BAA23644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892693 | Chr.15:50120328 - 50225890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map