Assay Details
Target Gene Details
Entrez Gene ID: | 57412 |
Gene Name: | arsenite methyltransferase |
Gene Aliases: |
CYT19 |
Location: |
Chr.10:102869453-102901899 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AS3MT | NM_020682.3 | 4 | 364 | NP_065733.2 |
| AF226730.1 | 4 | 301 | AAG09731.1 | |
| AK057833.1 | 4 | 383 | ||
| AK225972.1 | 4 | 383 | ||
| AK309737.1 | 4 | 296 | ||
| BC119637.1 | 4 | 268 | ||
| BC119638.2 | 4 | 268 | ||
| DA368970.1 | 4 | 364 |
Target Gene Details
Entrez Gene ID: | 100528007 |
Gene Name: | BORCS7-ASMT readthrough (NMD candidate) |
Gene Aliases: |
C10orf32-AS3MT, C10orf32-ASMT |
Location: |
Chr.10:102854210-102901899 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS7-ASMT | NR_037644.1 | 8 | 629 |
More Information
Set Membership: |
|

Genomic Map