Assay Details
Target Gene Details
Entrez Gene ID: | 56889 |
Gene Name: | transmembrane 9 superfamily member 3 |
Gene Aliases: |
EP70-P-iso, SMBP |
Location: |
Chr.10:96518110-96587052 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM9SF3 | NM_020123.3 | 15 | 3036 | NP_064508.3 |
| XM_011539976.2 | 15 | 3116 | XP_011538278.1 | |
| XM_011539977.1 | 15 | 3165 | XP_011538279.1 | |
| AF160213.1 | 15 | 2829 | AAF67014.1 | |
| AF269150.1 | 15 | 3036 | AAF98159.1 | |
| AK000756.1 | 13 | 2439 | BAA91362.1 | |
| AK027438.1 | 10 | 2086 | BAB55110.1 | |
| AK074824.1 | 12 | 2327 | BAC11232.1 | |
| AK075093.1 | 10 | 2074 | BAC11397.1 | |
| AK075451.1 | 4 | 1439 | ||
| AY358819.1 | 14 | 2706 | AAQ89178.1 | |
| BC020959.1 | 14 | 2616 | AAH20959.2 | |
| BC136788.1 | 15 | 3036 | ||
| BC136789.1 | 15 | 3036 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577627 | Chr.10:96515251 - 96710544 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map