Assay Details
Target Gene Details
Entrez Gene ID: | 9410 |
Gene Name: | small nuclear ribonucleoprotein U5 subunit 40 |
Gene Aliases: |
40K, HPRP8BP, PRP8BP, PRPF8BP, SPF38, WDR57 |
Location: |
Chr.1:31259568-31296797 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRNP40 | NM_004814.2 | 10 | 1114 | NP_004805.2 |
| AF090988.1 | 10 | 1083 | AAC69625.1 | |
| AK074516.1 | 10 | 1097 | ||
| AK123103.1 | 6 | 1524 | ||
| AK298823.1 | 11 | 1354 | ||
| AK313800.1 | ||||
| AL157420.1 | 4 | 719 | CAB75650.1 | |
| BC001494.2 | 10 | 1090 | AAH01494.1 | |
| EF011621.1 | 10 | 1089 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv161067 | Chr.1:31259127 - 31268295 on Build GRCh38 | Deletion |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map