Assay Details
Target Gene Details
Entrez Gene ID: | 84342 |
Gene Name: | component of oligomeric golgi complex 8 |
Gene Aliases: |
CDG2H, DOR1 |
Location: |
Chr.16:69328621-69339623 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG8 | NM_032382.4 | 4 | 1570 | NP_115758.3 |
| AK025968.1 | 2 | 692 | BAB15301.1 | |
| AK056344.1 | 4 | 1558 | BAB71157.1 | |
| AK225391.1 | 2 | 692 | ||
| AK302605.1 | 4 | 1583 | ||
| BC017492.1 | 2 | 374 | AAH17492.1 | |
| BC121022.1 | 4 | 1530 | ||
| BC121023.1 | 4 | 1530 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827715 | Chr.16:69311943 - 69340458 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map