Assay Details
Target Gene Details
Entrez Gene ID: | 29999 |
Gene Name: | fascin actin-bundling protein 3 |
Gene Aliases: |
- |
Location: |
Chr.7:127593635-127601797 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSCN3 | NM_020369.2 | NP_065102.1 | ||
| AF281049.1 | AAF91439.1 | |||
| AK225867.1 | ||||
| AK300518.1 | ||||
| DQ655930.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831122 | Chr.7:127503153 - 127717831 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map