Assay Details
Target Gene Details
Entrez Gene ID: | 51330 |
Gene Name: | TNF receptor superfamily member 12A |
Gene Aliases: |
CD266, FN14, TWEAKR |
Location: |
Chr.16:3020312-3022383 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF12A | NM_016639.2 | 4 | 549 | NP_057723.1 |
| AB035480.1 | 4 | 502 | BAA94792.1 | |
| AB035481.1 | 3 | 397 | BAB17850.1 | |
| AF191148.1 | 4 | 492 | AAF69108.1 | |
| BC002718.2 | 4 | 520 | AAH02718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637645 | Chr.16:3021600 - 3057596 on Build GRCh38 | Gain |
|
| nsv509588 | Chr.16:2919880 - 3043776 on Build GRCh38 | Insertion |
|
| nsv833125 | Chr.16:3017124 - 3131817 on Build GRCh38 | Loss |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map