Assay Details
Target Gene Details
Entrez Gene ID: | 53407 |
Gene Name: | syntaxin 18 |
Gene Aliases: |
Ufe1 |
Location: |
Chr.4:4418968-4542338 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX18 | NM_016930.2 | 11 | 1199 | NP_058626.1 |
| XM_011513480.1 | 12 | 1271 | XP_011511782.1 | |
| XM_011513483.1 | 11 | 1172 | XP_011511785.1 | |
| XM_011513485.2 | 11 | 1126 | XP_011511787.1 | |
| XM_017008297.1 | 12 | 1852 | XP_016863786.1 | |
| AB028741.1 | 11 | 1118 | BAA95213.1 | |
| AK310026.1 | ||||
| AL832867.1 | 5 | 465 | ||
| BC014613.2 | 11 | 1121 | AAH14613.1 | |
| BX647083.1 | 3 | 2538 |
More Information
Set Membership: |
|

Genomic Map