Assay Details
Target Gene Details
Entrez Gene ID: | 101928402 |
Gene Name: | uncharacterized LOC101928402 |
Gene Aliases: |
- |
Location: |
Chr.X:123959233-123961341 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928402 | NR_130770.1 | |||
| AI633959.1 | ||||
| DW416112.1 |
Target Gene Details
Entrez Gene ID: | 10735 |
Gene Name: | stromal antigen 2 |
Gene Aliases: |
SA-2, SA2, SCC3B, bA517O1.1 |
Location: |
Chr.X:123960560-124102656 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAG2 | NM_001042749.2 | NP_001036214.1 | ||
| XM_005262357.2 | XP_005262414.1 | |||
| XM_005262361.2 | XP_005262418.1 | |||
| XM_017029235.1 | XP_016884724.1 | |||
| AK098737.1 | ||||
| AK298633.1 | ||||
| AK313559.1 | ||||
| BQ425643.1 | ||||
| BX640970.1 | CAE45985.1 | |||
| DA534452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758890 | Chr.X:123830966 - 124028715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map