Assay Details
Target Gene Details
Entrez Gene ID: | 60561 |
Gene Name: | RAD50 interactor 1 |
Gene Aliases: |
RINT-1 |
Location: |
Chr.7:105532085-105567688 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RINT1 | NM_021930.4 | NP_068749.3 | ||
| XM_005250524.3 | XP_005250581.1 | |||
| XM_011516458.2 | XP_011514760.1 | |||
| XM_017012496.1 | XP_016867985.1 | |||
| XM_017012497.1 | XP_016867986.1 | |||
| XM_017012498.1 | XP_016867987.1 | |||
| AF317622.1 | AAG42101.1 | |||
| AK021847.1 | BAB13910.1 | |||
| AK055508.1 | ||||
| AK057757.1 | ||||
| AK308126.1 | ||||
| AL832825.1 | ||||
| BC068483.1 | AAH68483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464668 | Chr.7:105448246 - 105568045 on Build GRCh38 | Gain |
|
| esv2759549 | Chr.7:105117803 - 105568045 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map