Assay Details
Target Gene Details
Entrez Gene ID: | 122651 |
Gene Name: | ribonuclease A family member 11 (inactive) |
Gene Aliases: |
C14orf6, HEL-S-84p, RAJ1 |
Location: |
Chr.14:20582892-20590258 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE11 | NM_145250.3 | 3 | 1047 | NP_660293.1 |
| AK292348.1 | 2 | 1048 | ||
| BX161458.1 | 3 | 1019 | CAD61920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map