Assay Details
Target Gene Details
Entrez Gene ID: | 51367 |
Gene Name: | POP5 homolog, ribonuclease P/MRP subunit |
Gene Aliases: |
HSPC004, RPP2, RPP20, hPop5 |
Location: |
Chr.12:120579045-120581452 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POP5 | NM_015918.3 | 5 | 500 | NP_057002.2 |
| NM_198202.1 | 4 | 354 | NP_937845.1 | |
| XM_011538441.1 | 5 | 612 | XP_011536743.1 | |
| AA918619.1 | 1 | 296 | ||
| AF070660.1 | 5 | 484 | AAD20966.1 | |
| AF117232.1 | 5 | 484 | AAF17213.1 | |
| AJ306296.1 | 5 | 487 | CAC59822.1 | |
| AK223206.1 | 5 | 472 | BAD96926.1 | |
| AK303144.1 | 4 | 335 | ||
| BC012505.1 | 5 | 487 | AAH12505.1 | |
| BI668578.1 | 5 | 507 | ||
| CB995722.1 | 5 | 622 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv906 | Chr.12:120535880 - 120580638 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map