Assay Details
Target Gene Details
Entrez Gene ID: | 732 |
Gene Name: | complement component 8, beta polypeptide |
Gene Aliases: |
C82 |
Location: |
Chr.1:56929210-56974383 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8B | NM_000066.3 | 12 | 1896 | NP_000057.2 |
| NM_001278543.1 | 13 | 2118 | NP_001265472.1 | |
| NM_001278544.1 | 13 | 2025 | NP_001265473.1 | |
| XM_017002235.1 | 13 | 1964 | XP_016857724.1 | |
| AK292740.1 | 12 | 1751 | ||
| AK298425.1 | 13 | 1942 | ||
| AK298444.1 | 13 | 2118 | ||
| AK313382.1 | 12 | 1768 | ||
| AK316209.1 | 13 | 2115 | ||
| BC130575.1 | 12 | 1729 | ||
| CD364465.1 | 1 | 283 | ||
| M16973.1 | 12 | 1731 | AAA51862.1 | |
| X04393.1 | 11 | 1566 | CAA27981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2557221 | Chr.1:56928558 - 56929482 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map