Assay Details
Target Gene Details
Entrez Gene ID: | 9332 |
Gene Name: | CD163 molecule |
Gene Aliases: |
M130, MM130, SCARI1 |
Location: |
Chr.12:7470816-7503818 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD163 | NM_004244.5 | NP_004235.4 | ||
| NM_203416.3 | NP_981961.2 | |||
| XM_005253528.4 | 7 | 1941 | XP_005253585.1 | |
| XM_005253529.4 | XP_005253586.1 | |||
| XM_017020231.1 | XP_016875720.1 | |||
| AK226017.1 | ||||
| BC051281.1 | AAH51281.1 | |||
| DQ058615.1 | AAY99762.1 | |||
| Z22968.1 | CAA80541.1 | |||
| Z22969.1 | CAA80542.1 | |||
| Z22970.1 | CAA80543.1 | |||
| Z22971.1 | 7 | 1909 | CAA80544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557269 | Chr.12:7471128 - 7538538 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map