Assay Details
Target Gene Details
Entrez Gene ID: | 7401 |
Gene Name: | clarin 1 |
Gene Aliases: |
RP61, USH3, USH3A |
Location: |
Chr.3:150926163-150972999 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLRN1 | NM_001195794.1 | 1 | 246 | NP_001182723.1 |
| NM_001256819.1 | 1 | 246 | NP_001243748.1 | |
| NM_174878.2 | 1 | 246 | NP_777367.1 | |
| NR_046380.2 | 1 | 246 | ||
| AF482697.1 | 1 | 247 | AAN07148.1 | |
| AF495717.1 | AAM88774.1 | |||
| BC074970.2 | AAH74970.1 | |||
| BC074971.2 | AAH74971.1 |
Target Gene Details
Entrez Gene ID: | 116933 |
Gene Name: | CLRN1 antisense RNA 1 |
Gene Aliases: |
CLRN1OS, UCRP |
Location: |
Chr.3:150972678-151079830 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLRN1-AS1 | NR_024066.1 | 1 | 77 | |
| AF388367.1 | 1 | 77 | ||
| AK126442.1 |
More Information
Set Membership: |
|

Genomic Map