Assay Details
Target Gene Details
Entrez Gene ID: | 81890 |
Gene Name: | queuine tRNA-ribosyltransferase catalytic subunit 1 |
Gene Aliases: |
FP3235, TGT, TGUT |
Location: |
Chr.19:10701436-10713367 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QTRT1 | NM_031209.2 | NP_112486.1 | ||
| AF302783.1 | AAG60033.1 | |||
| AF302784.1 | AAG60034.1 | |||
| AF370379.1 | AAQ15215.1 | |||
| AK057877.1 | ||||
| AK225173.1 | ||||
| AK294166.1 | ||||
| AK314308.1 | ||||
| BC015350.1 | AAH15350.1 | |||
| CT003728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
| nsv833746 | Chr.19:10678751 - 10842756 on Build GRCh38 | Loss |
|
| nsv1160585 | Chr.19:10674243 - 10771056 on Build GRCh38 | Deletion |
|
| nsv1061882 | Chr.19:10704200 - 10737778 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map