Assay Details
Target Gene Details
Entrez Gene ID: | 160760 |
Gene Name: | PTC7 protein phosphatase homolog |
Gene Aliases: |
TA-PP2C, TAPP2C |
Location: |
Chr.12:110534432-110583259 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPTC7 | NM_139283.1 | NP_644812.1 | ||
| AF385435.1 | AAM43836.1 | |||
| AK124744.1 | ||||
| AY357944.1 | AAQ57274.1 | |||
| BC111551.1 | AAI11552.1 | |||
| CR749216.1 | CAH18073.1 |
More Information
Set Membership: |
|

Genomic Map