Assay Details
Target Gene Details
Entrez Gene ID: | 89848 |
Gene Name: | FCH and double SH3 domains 1 |
Gene Aliases: |
NWK2 |
Location: |
Chr.5:141639302-141651453 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCHSD1 | NM_033449.2 | 20 | 2827 | NP_258260.1 |
| AK000007.1 | 20 | 2811 | ||
| AK027281.1 | 1 | 818 | ||
| AK126203.1 | 2 | 1689 | ||
| AY217346.1 | 20 | 2827 | AAO45099.1 | |
| AY358478.1 | 11 | 2445 | AAQ88842.1 |
Target Gene Details
Entrez Gene ID: | 285613 |
Gene Name: | RELT like 2 |
Gene Aliases: |
C5orf16 |
Location: |
Chr.5:141636946-141641077 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RELL2 | NM_001130029.1 | 8 | 1886 | NP_001123501.1 |
| NM_173828.4 | 7 | 2314 | NP_776189.3 | |
| XM_005268414.3 | 6 | 2231 | XP_005268471.1 | |
| XM_011537624.1 | 6 | 2247 | XP_011535926.1 | |
| XM_011537625.2 | 6 | 2434 | XP_011535927.1 | |
| AK054889.1 | 5 | 2061 | ||
| AK075064.1 | 7 | 1141 | BAC11380.1 | |
| AY358163.1 | 7 | 2250 | AAQ88530.1 | |
| BC063469.1 | 8 | 1355 | AAH63469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map