Assay Details
Target Gene Details
Entrez Gene ID: | 54790 |
Gene Name: | tet methylcytosine dioxygenase 2 |
Gene Aliases: |
KIAA1546, MDS |
Location: |
Chr.4:105145875-105279803 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TET2 | NM_001127208.2 | NP_001120680.1 | ||
| NM_017628.4 | NP_060098.3 | |||
| XM_005263082.2 | XP_005263139.1 | |||
| XM_006714242.3 | XP_006714305.1 | |||
| XM_017008319.1 | XP_016863808.1 | |||
| AI084057.1 | ||||
| AK055149.1 | ||||
| BC150180.1 | ||||
| BC150201.1 | ||||
| BX640738.1 | CAE45851.1 | |||
| DA391215.1 | ||||
| DR005542.1 | ||||
| FM992369.1 |
Target Gene Details
Entrez Gene ID: | 104384744 |
Gene Name: | TET2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:105171354-105352881 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TET2-AS1 | NR_126420.1 | |||
| AI401598.1 | ||||
| BX100522.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830025 | Chr.4:105067719 - 105216789 on Build GRCh38 | Loss |
|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
| esv3601659 | Chr.4:105145497 - 105197242 on Build GRCh38 | Loss |
|
| nsv595007 | Chr.4:104948391 - 105445052 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map