Assay Details
Target Gene Details
Entrez Gene ID: | 79637 |
Gene Name: | armadillo repeat containing 7 |
Gene Aliases: |
- |
Location: |
Chr.17:75109952-75130265 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC7 | NM_001304271.1 | NP_001291200.1 | ||
| NM_001304272.1 | NP_001291201.1 | |||
| NM_001304273.1 | NP_001291202.1 | |||
| NM_024585.3 | NP_078861.1 | |||
| AK025813.1 | BAB15242.1 | |||
| AK300997.1 | ||||
| AK310697.1 | ||||
| AW081695.1 | ||||
| BC011728.1 | AAH11728.1 | |||
| BI826938.1 | ||||
| CR748475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641258 | Chr.17:75107625 - 75120521 on Build GRCh38 | Loss |
|
| nsv470602 | Chr.17:75090871 - 75208042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map