Assay Details
Target Gene Details
Entrez Gene ID: | 8437 |
Gene Name: | RAS protein activator like 1 |
Gene Aliases: |
RASAL |
Location: |
Chr.12:113096515-113136239 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASAL1 | NM_001193520.1 | 22 | 2806 | NP_001180449.1 |
| NM_001193521.1 | 21 | 2707 | NP_001180450.1 | |
| NM_001301202.1 | 21 | 2639 | NP_001288131.1 | |
| NM_004658.2 | 22 | 2815 | NP_004649.2 | |
| XM_005253950.4 | 22 | 2821 | XP_005254007.1 | |
| XM_006719641.3 | 22 | 2660 | XP_006719704.1 | |
| XM_006719642.3 | 21 | 2790 | XP_006719705.1 | |
| XM_011538852.2 | 21 | 2793 | XP_011537154.1 | |
| XM_011538853.2 | 21 | 2787 | XP_011537155.1 | |
| XM_011538854.2 | 11 | 1533 | XP_011537156.1 | |
| XM_017020028.1 | 22 | 2818 | XP_016875517.1 | |
| XM_017020029.1 | 22 | 2903 | XP_016875518.1 | |
| XM_017020030.1 | 23 | 2931 | XP_016875519.1 | |
| XM_017020031.1 | 23 | 2928 | XP_016875520.1 | |
| AB208935.1 | 22 | 2791 | BAD92172.1 | |
| AF086713.1 | 22 | 2792 | AAD09006.1 | |
| AK294350.1 | ||||
| BC014420.1 | 21 | 2598 | AAH14420.1 | |
| BC093724.1 | AAH93724.1 | |||
| BC143261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560265 | Chr.12:113084969 - 113113365 on Build GRCh38 | Loss |
|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| nsv560264 | Chr.12:113050137 - 113136469 on Build GRCh38 | Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map