Assay Details
Target Gene Details
Entrez Gene ID: | 29087 |
Gene Name: | thymocyte nuclear protein 1 |
Gene Aliases: |
HSPC144, MDS012, MY105, THY28, THY28KD |
Location: |
Chr.11:134248278-134253401 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THYN1 | NM_001037304.1 | NP_001032381.1 | ||
| NM_001037305.1 | NP_001032382.1 | |||
| NM_014174.2 | NP_054893.1 | |||
| NM_199297.1 | NP_954994.1 | |||
| NM_199298.1 | NP_954995.1 | |||
| XM_005271527.3 | XP_005271584.1 | |||
| XM_005271528.3 | XP_005271585.1 | |||
| AF059619.1 | AAG43118.1 | |||
| AF087886.1 | AAP97185.1 | |||
| AF161493.1 | AAF29108.1 | |||
| AF182413.1 | AAG14949.1 | |||
| AK308498.1 | 5 | 825 | ||
| AK311024.1 | 5 | 928 | ||
| BC006978.1 | AAH06978.1 | |||
| BC093074.1 | AAH93074.1 | |||
| BQ640137.1 | ||||
| BU685681.1 | ||||
| BU934600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain |
|
| nsv832299 | Chr.11:134170258 - 134346785 on Build GRCh38 | Loss |
|

Genomic Map