Assay Details
Target Gene Details
Entrez Gene ID: | 3748 |
Gene Name: | potassium voltage-gated channel subfamily C member 3 |
Gene Aliases: |
KSHIIID, KV3.3, SCA13 |
Location: |
Chr.19:50311942-50333515 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNC3 | NM_004977.2 | 2 | 2013 | NP_004968.2 |
| NR_110912.1 | ||||
| XM_006723203.2 | 2 | 1718 | XP_006723266.1 | |
| XM_011526925.2 | 2 | 1718 | XP_011525227.1 | |
| XM_011526926.1 | 2 | 1718 | XP_011525228.1 | |
| XM_011526928.2 | 2 | 1718 | XP_011525230.1 | |
| AB208930.1 | 1 | 819 | BAD92167.1 | |
| AF055989.1 | 2 | 2013 | AAC24118.1 | |
| AK127492.1 | BAC87004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953603 | Chr.19:50311544 - 50335643 on Build GRCh38 | Deletion |
|
| nsv1066389 | Chr.19:50319151 - 50453251 on Build GRCh38 | Gain |
|
| nsv523948 | Chr.19:50178521 - 50354566 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map