Assay Details
Target Gene Details
Entrez Gene ID: | 3846 |
Gene Name: | keratin associated protein 5-9 |
Gene Aliases: |
KRN1, KRTAP5-1, KRTAP5.9 |
Location: |
Chr.11:71548420-71549607 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP5-9 | NM_005553.3 | 1 | 122 | NP_005544.4 |
| BC101744.1 | ||||
| X63755.1 | 1 | 122 | CAA45283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626782 | Chr.11:71538088 - 71562989 on Build GRCh38 | Gain |
|
| esv2744750 | Chr.11:71526540 - 71564429 on Build GRCh38 | Deletion |
|
| nsv555402 | Chr.11:71489274 - 71578043 on Build GRCh38 | Gain |
|
| esv3626781 | Chr.11:71526772 - 71565036 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map