Assay Details
Target Gene Details
Entrez Gene ID: | 284306 |
Gene Name: | zinc finger protein 547 |
Gene Aliases: |
- |
Location: |
Chr.19:57363435-57379559 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF547 | NM_173631.3 | 4 | 660 | NP_775902.2 |
| AK055662.1 | 4 | 572 | BAB70979.1 | |
| AK291464.1 | 4 | 507 | ||
| AL832545.1 | 4 | 528 | ||
| BC042681.1 | 4 | 529 | AAH42681.2 |
More Information
Set Membership: |
|

Genomic Map