Assay Details
Target Gene Details
Entrez Gene ID: | 10241 |
Gene Name: | calcium binding and coiled-coil domain 2 |
Gene Aliases: |
NDP52 |
Location: |
Chr.17:48830988-48865245 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALCOCO2 | NM_001261390.1 | 2 | 197 | NP_001248319.1 |
| NM_001261391.1 | 2 | 197 | NP_001248320.1 | |
| NM_001261393.1 | 2 | 197 | NP_001248322.1 | |
| NM_001261395.1 | NP_001248324.1 | |||
| NM_005831.4 | 2 | 197 | NP_005822.1 | |
| AK222666.1 | 2 | 147 | BAD96386.1 | |
| AK223227.1 | 2 | 140 | BAD96947.1 | |
| AK293137.1 | ||||
| AK293329.1 | 2 | 159 | ||
| AK298177.1 | 2 | 140 | ||
| AK303313.1 | 2 | 150 | ||
| AK304024.1 | 2 | 150 | ||
| AK310648.1 | 2 | 150 | ||
| AK314796.1 | 2 | 150 | ||
| AL042638.1 | 2 | 195 | ||
| AL046722.1 | 2 | 197 | ||
| BC004130.2 | 2 | 136 | AAH04130.1 | |
| BC015893.2 | 1 | 106 | AAH15893.1 | |
| BG773301.1 | 2 | 179 | ||
| CR456763.1 | 1 | 96 | CAG33044.1 | |
| U22897.1 | 2 | 150 | AAA75297.1 |
More Information
Set Membership: |
|

Genomic Map