Assay Details
Target Gene Details
Entrez Gene ID: | 100499483 |
Gene Name: | coiled-coil domain containing 180 |
Gene Aliases: |
BDAG1, C9orf174, KIAA1529 |
Location: |
Chr.9:97307628-97377295 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC180 | NM_020893.2 | 14 | 1752 | NP_065944.2 |
| AK302965.1 | 15 | 1826 | ||
| BC047097.1 | 15 | 1695 | AAH47097.2 | |
| CR749320.1 | 14 | 1694 | CAH18175.1 |
Target Gene Details
Entrez Gene ID: | 57653 |
Gene Name: | LOC100499484-C9orf174 readthrough |
Gene Aliases: |
- |
Location: |
Chr.9:97238426-97377295 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100499484-C9ORF174 | NR_036527.1 | 28 | 3047 | |
| NR_036528.1 | 28 | 3047 | ||
| NR_036529.1 | 25 | 2607 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762823 | Chr.9:97321590 - 97343500 on Build GRCh38 | Loss |
|
| esv3621188 | Chr.9:97318756 - 97341085 on Build GRCh38 | Loss |
|

Genomic Map