Assay Details
Target Gene Details
Entrez Gene ID: | 5567 |
Gene Name: | protein kinase cAMP-activated catalytic subunit beta |
Gene Aliases: |
PKA C-beta, PKACB |
Location: |
Chr.1:84077975-84238498 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKACB | NM_002731.3 | 1 | 278 | NP_002722.1 |
| NM_207578.2 | 1 | 278 | NP_997461.1 | |
| XM_017001713.1 | 1 | 220 | XP_016857202.1 | |
| AK074904.1 | 1 | 300 | ||
| AK314550.1 | 1 | 174 | ||
| BC016285.1 | 1 | 109 | AAH16285.1 | |
| BC035058.1 | 1 | 196 | AAH35058.1 | |
| HY160843.1 | 1 | 278 | ||
| M34181.1 | AAA60170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1732 | Chr.1:84033415 - 84078952 on Build GRCh38 | Deletion |
|
| nsv546689 | Chr.1:84065434 - 84205133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map