Assay Details
Target Gene Details
Entrez Gene ID: | 8508 |
Gene Name: | nipsnap homolog 1 (C. elegans) |
Gene Aliases: |
- |
Location: |
Chr.22:29554808-29581337 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIPSNAP1 | NM_001202502.1 | NP_001189431.1 | ||
| NM_003634.3 | NP_003625.2 | |||
| AJ001258.1 | CAA04632.1 | |||
| AK098367.1 | ||||
| AK290953.1 | ||||
| AK298816.1 | ||||
| AK314407.1 | ||||
| BC002371.2 | AAH02371.1 | |||
| BC006473.1 | AAH06473.1 | |||
| DA133702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834172 | Chr.22:29503047 - 29661959 on Build GRCh38 | Loss |
|
| esv3647539 | Chr.22:29545022 - 29571563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map