Assay Details
Target Gene Details
Entrez Gene ID: | 132321 |
Gene Name: | chromosome 4 open reading frame 33 |
Gene Aliases: |
- |
Location: |
Chr.4:129093674-129112688 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf33 | NM_173487.2 | NP_775758.2 | ||
| AK091022.1 | BAC03568.1 | |||
| BP871612.1 |
Target Gene Details
Entrez Gene ID: | 132320 |
Gene Name: | sodium channel and clathrin linker 1 |
Gene Aliases: |
CAP-1A, CAP1A |
Location: |
Chr.4:128875206-129093607 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCLT1 | NM_001300897.1 | NP_001287826.1 | ||
| NM_001300898.1 | NP_001287827.1 | |||
| NM_144643.3 | NP_653244.2 | |||
| XM_017007716.1 | XP_016863205.1 | |||
| XM_017007717.1 | XP_016863206.1 | |||
| XM_017007718.1 | XP_016863207.1 | |||
| AK055217.1 | BAB70876.1 | |||
| CD691962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822718 | Chr.4:129093216 - 129093671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map