Assay Details
Target Gene Details
Entrez Gene ID: | 55088 |
Gene Name: | coiled-coil domain containing 186 |
Gene Aliases: |
C10orf118 |
Location: |
Chr.10:114120862-114174312 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC186 | NM_001321829.1 | 17 | 5476 | NP_001308758.1 |
| NM_018017.3 | 16 | 5351 | NP_060487.2 | |
| NR_135815.1 | 16 | 5963 | ||
| XM_011539915.2 | 15 | 4694 | XP_011538217.1 | |
| AK094498.1 | 1 | 838 | ||
| AL834368.1 | 15 | 4551 | CAD39031.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052267 | Chr.10:114103969 - 114155081 on Build GRCh38 | Loss |
|
| dgv287e212 | Chr.10:114092418 - 114127845 on Build GRCh38 | Loss |
|
| dgv288e212 | Chr.10:114103827 - 114127864 on Build GRCh38 | Loss |
|

Genomic Map