Assay Details
Target Gene Details
Entrez Gene ID: | 55794 |
Gene Name: | DEAD-box helicase 28 |
Gene Aliases: |
MDDX28 |
Location: |
Chr.16:68021274-68023867 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX28 | NM_018380.3 | 1 | 1431 | NP_060850.2 |
| AK002144.1 | 1 | 1006 | BAA92106.1 | |
| BC024273.1 | 1 | 773 | AAH24273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2666619 | Chr.16:68003989 - 68045407 on Build GRCh38 | Deletion |
|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| esv3638897 | Chr.16:68004075 - 68045657 on Build GRCh38 | Loss |
|
| esv3638899 | Chr.16:68008886 - 68035332 on Build GRCh38 | Loss |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map