Assay Details
Target Gene Details
Entrez Gene ID: | 6480 |
Gene Name: | ST6 beta-galactoside alpha-2,6-sialyltransferase 1 |
Gene Aliases: |
SIAT1, ST6GalI, ST6N |
Location: |
Chr.3:186930343-187078553 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GAL1 | NM_173216.2 | 2 | 358 | NP_775323.1 |
| NM_173217.2 | 2 | 358 | NP_775324.1 | |
| XM_006713734.1 | 3 | 613 | XP_006713797.1 | |
| XM_011513085.1 | 3 | 621 | XP_011511387.1 | |
| XM_017007065.1 | 4 | 705 | XP_016862554.1 | |
| A17362.1 | 2 | 163 | CAA01327.1 | |
| DB152155.1 | 2 | 399 | ||
| DB235666.1 | 2 | 147 | ||
| X17247.1 | 2 | 157 | CAA35111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001111 | Chr.3:186796249 - 187014891 on Build GRCh38 | Gain |
|
| esv34136 | Chr.3:186697095 - 187036942 on Build GRCh38 | Loss |
|
| nsv1003732 | Chr.3:186888059 - 186967511 on Build GRCh38 | Gain |
|
| nsv508975 | Chr.3:186936621 - 186986854 on Build GRCh38 | Insertion |
|
| nsv1004008 | Chr.3:186879814 - 187204053 on Build GRCh38 | Gain |
|
| esv2759207 | Chr.3:186891945 - 187066496 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs71634020] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map