Assay Details
Target Gene Details
Entrez Gene ID: | 128646 |
Gene Name: | signal regulatory protein delta |
Gene Aliases: |
PTPNS1L2, dJ576H24.4 |
Location: |
Chr.20:1534251-1557697 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPD | NM_178460.2 | 2 | 247 | NP_848555.2 |
| AK093083.1 | 2 | 1393 | ||
| BC033502.1 | 2 | 247 | AAH33502.1 | |
| DA466740.1 | 3 | 447 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| nsv510779 | Chr.20:1528684 - 1633316 on Build GRCh38 | Deletion |
|
| esv2758777 | Chr.20:1496135 - 1698610 on Build GRCh38 | Gain+Loss |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| nsv833894 | Chr.20:1538787 - 1772720 on Build GRCh38 | Gain |
|
| esv3644980 | Chr.20:1509522 - 1569123 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map