Assay Details
Target Gene Details
Entrez Gene ID: | 5166 |
Gene Name: | pyruvate dehydrogenase kinase 4 |
Gene Aliases: |
- |
Location: |
Chr.7:95583497-95596613 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDK4 | NM_002612.3 | 11 | 1539 | NP_002603.1 |
| AK096428.1 | 12 | 1338 | ||
| AK098151.1 | 7 | 853 | ||
| AK223328.1 | 11 | 1435 | BAD97048.1 | |
| AK312424.1 | ||||
| BC040239.1 | 11 | 1419 | AAH40239.1 | |
| U54617.1 | 11 | 1441 | AAC50669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map