Assay Details
Target Gene Details
Entrez Gene ID: | 80341 |
Gene Name: | BPI fold containing family B member 2 |
Gene Aliases: |
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2 |
Location: |
Chr.20:33007578-33023709 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFB2 | NM_025227.2 | NP_079503.1 | ||
| AF465765.1 | AAM73983.1 | |||
| AK027068.1 | ||||
| AK172819.1 | BAD18788.1 | |||
| AK304064.1 | ||||
| AY358723.1 | AAQ89085.1 | |||
| BC034415.1 | AAH34415.1 | |||
| BP333980.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2758792 | Chr.20:32908001 - 33160360 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map