Assay Details
Target Gene Details
Entrez Gene ID: | 2914 |
Gene Name: | glutamate metabotropic receptor 4 |
Gene Aliases: |
GPRC1D, MGLUR4, mGlu4 |
Location: |
Chr.6:34018643-34155622 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM4 | NM_000841.3 | 11 | 3361 | NP_000832.1 |
| NM_001256809.2 | 9 | 2744 | NP_001243738.1 | |
| NM_001256811.2 | 10 | 3220 | NP_001243740.1 | |
| NM_001256812.2 | 10 | 2445 | NP_001243741.1 | |
| NM_001256813.2 | 11 | 2669 | NP_001243742.1 | |
| NM_001282847.1 | 10 | 3144 | NP_001269776.1 | |
| XM_017010790.1 | 11 | 3299 | XP_016866279.1 | |
| XM_017010791.1 | 11 | 3354 | XP_016866280.1 | |
| XM_017010792.1 | 10 | 2898 | XP_016866281.1 | |
| XM_017010793.1 | 10 | 2649 | XP_016866282.1 | |
| AA993227.1 | ||||
| AK122836.1 | 8 | 3335 | ||
| AK122982.1 | 11 | 2654 | ||
| AK126746.1 | 10 | 2909 | ||
| AK131536.1 | 9 | 2744 | BAD18673.1 | |
| AK289534.1 | 11 | 3304 | ||
| AK289563.1 | 9 | 2218 | ||
| AK293913.1 | 10 | 2445 | ||
| AK293949.1 | 10 | 2471 | ||
| BC130526.1 | 10 | 3019 | ||
| BC130528.1 | 10 | 3019 | ||
| BC144057.1 | 9 | 2878 | ||
| U92457.1 | 10 | 2885 | AAB51762.1 | |
| X80818.1 | 10 | 3030 | CAA56784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| nsv602861 | Chr.6:33985869 - 34083140 on Build GRCh38 | Loss |
|
| nsv602843 | Chr.6:33964665 - 34227234 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map