Assay Details
Target Gene Details
Entrez Gene ID: | 7581 |
Gene Name: | zinc finger protein 33A |
Gene Aliases: |
KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPK |
Location: |
Chr.10:38010483-38067834 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF33A | NM_001278170.1 | 1 | 32 | NP_001265099.1 |
| NM_001278171.1 | 1 | 32 | NP_001265100.1 | |
| NM_001278173.1 | 1 | 32 | NP_001265102.1 | |
| NM_001278174.1 | 1 | 32 | NP_001265103.1 | |
| NM_001278175.1 | 1 | 32 | NP_001265104.1 | |
| NM_001278176.1 | 1 | 32 | NP_001265105.1 | |
| NM_001278177.1 | 1 | 32 | NP_001265106.1 | |
| NM_001278178.1 | 1 | 32 | NP_001265107.1 | |
| NM_001278179.1 | 1 | 32 | NP_001265108.1 | |
| NM_001324175.1 | 1 | 32 | NP_001311104.1 | |
| NM_001324176.1 | 1 | 32 | NP_001311105.1 | |
| NM_001324177.1 | 1 | 32 | NP_001311106.1 | |
| NM_001324178.1 | 1 | 32 | NP_001311107.1 | |
| NM_006954.1 | 1 | 32 | NP_008885.1 | |
| NM_006974.2 | 1 | 32 | NP_008905.1 | |
| NR_136709.1 | 1 | 32 | ||
| NR_136710.1 | 1 | 32 | ||
| XM_011519650.2 | XP_011517952.1 | |||
| XM_011519651.2 | XP_011517953.1 | |||
| XM_017016617.1 | XP_016872106.1 | |||
| AK027057.1 | BAB15642.1 | |||
| AK292844.1 | ||||
| AK303445.1 | ||||
| AL712670.1 | ||||
| BC017760.1 | ||||
| BF435065.1 | ||||
| D31763.1 | BAA06541.1 | |||
| DA276183.1 | ||||
| DA757208.1 | ||||
| DB134021.1 | ||||
| DB234051.1 | ||||
| DB235703.1 | 1 | 33 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040704 | Chr.10:37651096 - 38162989 on Build GRCh38 | Gain |
|
| esv3576438 | Chr.10:38002320 - 38607542 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map