Assay Details
Target Gene Details
Entrez Gene ID: | 6478 |
Gene Name: | siah E3 ubiquitin protein ligase 2 |
Gene Aliases: |
hSiah2 |
Location: |
Chr.3:150741123-150763476 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIAH2 | NM_005067.5 | 2 | 2355 | NP_005058.3 |
| AK055290.1 | 1 | 1801 | ||
| BC013082.2 | 2 | 2057 | AAH13082.1 | |
| BM970396.1 | 1 | 281 | ||
| U76248.1 | AAC51908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4061 | Chr.3:150720633 - 150752549 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map